2-Bromobenzyl alcohol ≥99.0%

Supplier: TCI America

Synonyms: (2-Bromophenyl)methanol

B1294-100G B1294-25G
TCB1294-100GEA 308.1 CAD
TCB1294-100G TCB1294-025G
2-Bromobenzyl alcohol ≥99.0%
2-Bromobenzyl alcohol

CAS Number: 18982-54-2
MDL Number: MFCD00004600
Molecular Formula: C7H7BrO
Molecular Weight: 187.04
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 80

Formula: C₇H₇BrO
MW: 187.04 g/mol
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: 80 °C
MDL Number: MFCD00004600
CAS Number: 18982-54-2
EINECS: 242-719-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2022e5ffccfabf9affb6fbdeb042d91606fd13df&vid=2022e5ffccfabf9affb6fbdeb042d91606fd13df&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR