3-Bromobenzyl alcohol ≥98.0%

Supplier: TCI America

B1287-5G B1287-25G
TCB1287-005GEA 53.72 CAD
TCB1287-005G TCB1287-025G
3-Bromobenzyl alcohol ≥98.0%
3-Bromobenzyl alcohol

CAS Number: 15852-73-0
MDL Number: MFCD00004629
Molecular Formula: C7H7BrO
Molecular Weight: 187.04
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 165
Flash Point (°C): 110
Specific Gravity (20/20): 1.56

Formula: C₇H₇BrO
Boiling Pt: 165 °C
Density: 1.56 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00004629
CAS Number: 15852-73-0
EINECS: 239-975-4

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR