3-Bromo-o-xylene ≥98.0%

Supplier: TCI America
Warning

Synonyms: 1-Bromo-2,3-dimethylbenzene

B1282-5G B1282-25G
TCB1282-5GEA 36.34 CAD
TCB1282-5G TCB1282-025G
3-Bromo-o-xylene ≥98.0%
3-Bromo-o-xylene

CAS Number: 576-23-8
MDL Number: MFCD00000069
Molecular Formula: C8H9Br
Molecular Weight: 185.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 214
Flash Point (°C): 80
Specific Gravity (20/20): 1.38


Other Product Names / Synonyms: 1-Bromo-2,3-dimethylbenzene

Formula: C₈H₉Br
Boiling Pt: 214 °C
Density: 1.38 (20 °C)
Flash Pt: 80 °C
MDL Number: MFCD00000069
CAS Number: 576-23-8
EINECS: 209-398-2
UN: 1701
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb9ff111ad35e965b979f221d1125437d8c5ddc6&vid=715bf229a05d755b243c039e01eff3e66188f51a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR