2-Bromothiazole ≥98.0%

Supplier: TCI America

B1280-25G B1280-250G
TCB1280-025GEA 126.4 CAD
TCB1280-025G TCB1280-250G
2-Bromothiazole ≥98.0%
2-Bromothiazole

CAS Number: 3034-53-5
MDL Number: MFCD00005316
Molecular Formula: C3H2BrNS
Molecular Weight: 164.02
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 171
Flash Point (°C): 63
Specific Gravity (20/20): 1.85
Storage Temperature: <0°C


Other Product Names / Synonyms: 2-Thiazolyl Bromide

Formula: C₃H₂BrNS
Boiling Pt: 171 °C
Density: 1.85 (20 °C)
Flash Pt: 63 °C
MDL Number: MFCD00005316
CAS Number: 3034-53-5
EINECS: 221-229-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d587f4fec2fd07f6fa94ef62290868c5122e9f77&vid=d587f4fec2fd07f6fa94ef62290868c5122e9f77&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR