2,2'-Bithiophene ≥98.0%
Supplier: TCI America
Synonyms:
2,2′-Dithienyl, 2,2′-Bithienyl
B1276-25G
B1276-1G
TCB1276-25GEA
159.58
CAD
TCB1276-25G
TCB1276-001G
2,2'-Bithiophene ≥98.0%
2,2'-Bithiophene
CAS Number: 492-97-7
MDL Number: MFCD00005414
Molecular Formula: C8H6S2
Molecular Weight: 166.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 260
Flash Point (°C): 110
Freezing point (°C): 32
Product Categories: Materials Chemistry, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 2,2'-Bithienyl
Formula:
C₈H₆S₂ MW: 166.27 g/mol Boiling Pt: 260 °C Flash Pt: 110 °C |
MDL Number:
MFCD00005414 CAS Number: 492-97-7 EINECS: 207-767-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...