3-Brompropylamine hydrobromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 3-Bromopropan-1-amine hydrobromide, 3-Bromopropylammonium bromide

B1263-500G B1263-25G B1263-100G
TCB1263-500GEA 613.04 CAD
TCB1263-500G TCB1263-025G TCB1263-100G
3-Brompropylamine hydrobromide ≥98.0% (by titrimetric analysis)
3-Brompropylamine hydrobromide

CAS Number: 5003-71-4
MDL Number: MFCD00012912
Molecular Formula: C3H8BrN
Molecular Weight: 218.92
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 174


Other Product Names / Synonyms: 3-Aminopropyl Bromide Hydrobromide

Formula: BrH₂CCH₂CH₂NH₃Br
MW: 218.92 g/mol
Melting Pt: 174 °C
MDL Number: MFCD00012912
CAS Number: 5003-71-4
EINECS: 225-675-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aab9bb0f7da6b6725d00a9a7afd7c83aff8b0627&vid=aab9bb0f7da6b6725d00a9a7afd7c83aff8b0627&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR