2-Bromo-4-nitrotoluene ≥98.0%

Supplier: TCI America
Warning

B1262-25G B1262-250G
TCB1262-025GEA 88.48 CAD
TCB1262-025G TCB1262-250G
2-Bromo-4-nitrotoluene ≥98.0%
2-Bromo-4-nitrotoluene

CAS Number: 7745-93-9
MDL Number: MFCD00007195
Molecular Formula: C7H6BrNO2
Molecular Weight: 216.03
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 175
Melting point (°C): 77

Formula: C₇H₆BrNO₂
Boiling Pt: 175 °C
Melting Pt: 77 °C
MDL Number: MFCD00007195
CAS Number: 7745-93-9
EINECS: 231-809-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a625e98219f0ab4f3864d8410100a8d6de05d538&vid=93d88f92d28895c78a186ee104cc39043b0969bd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR