2-Bromomesitylene ≥99.0%

Supplier: TCI America

Synonyms: 2,4,6-Trimethylbromobenzene, 2-Bromo-1,3,5-trimethylbenzene, Mesityl bromide, 2-Bromomesitylene

B1261-25G B1261-500G B1261-100G
TCB1261-025GEA 59.39 CAD
TCB1261-025G TCB1261-500G TCB1261-100G
2-Bromomesitylene ≥99.0%
2-Bromomesitylene

CAS Number: 576-83-0
MDL Number: MFCD00000073
Molecular Formula: C9H11Br
Molecular Weight: 199.09
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 225
Flash Point (°C): 96
Specific Gravity (20/20): 1.33


Other Product Names / Synonyms: 2-Bromomesitylene, 2-Bromo-1,3,5-trimethylbenzene

Formula: C₉H₁₁Br
Boiling Pt: 225 °C
Melting Pt: 2 °C
Density: 1.33 (20 °C)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00000073
CAS Number: 576-83-0
EINECS: 209-405-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23d129b4354f8ab4493856146d4ce607bd245037&vid=901d3b2f341c4421c02f70ec1a74528880260653&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR