4-Bromoisophthalic acid ≥97.0%

Supplier: TCI America

B1260-10G B1260-25G
TCB1260-010GEA 303.36 CAD
TCB1260-010G TCB1260-25G
4-Bromoisophthalic acid ≥97.0%
4-Bromoisophthalic acid

CAS Number: 6939-93-1
MDL Number: MFCD00002404
Molecular Formula: C8H5BrO4
Molecular Weight: 245.03
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal

Formula: C₈H₅BrO₄
MW: 245.03 g/mol
Melting Pt: 303…304 °C
MDL Number: MFCD00002404
CAS Number: 6939-93-1
EINECS: 230-078-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e09ae9075cb3e7ea03a52a77cd1b0a73e386c05&vid=1e09ae9075cb3e7ea03a52a77cd1b0a73e386c05&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR