(3-Bromopropyl)benzene ≥97.0%
Supplier: TCI America
|
Warning
|
Synonyms:
1-Bromo-3-phenylpropane
B1237-500G
B1237-25G
B1237-100G
TCB1237-500GEA
329.38
CAD
TCB1237-500G
TCB1237-025G
TCB1237-100G
(3-Bromopropyl)benzene ≥97.0%
(3-Bromopropyl)benzene
CAS Number: 637-59-2
MDL Number: MFCD00000257
Molecular Formula: C9H11Br
Molecular Weight: 199.09
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 129
Flash Point (°C): 101
Specific Gravity (20/20): 1.32
Other Product Names / Synonyms: 1-Bromo-3-phenylpropane, (3-Bromopropyl)benzene
Formula:
BrH₂CCH₂CH₂(C₆H₅) MW: 199.09 g/mol Boiling Pt: 129 °C Density: 1.32 (20 °C) Flash Pt: 101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000257 CAS Number: 637-59-2 EINECS: 211-294-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...