1-Bromo-3-iodobenzene ≥97.0% stabilised

Supplier: TCI America
Warning

Synonyms: 3-Bromoiodobenzene

Stabilization: Stabilised with Stabilised

B1236-5G B1236-25G B1236-100G
TCB1236-005GEA 26.86 CAD
TCB1236-005G TCB1236-025G TCB1236-100G
1-Bromo-3-iodobenzene ≥97.0% stabilised
1-Bromo-3-iodobenzene

(stabilized with Copper chip)
CAS Number: 591-18-4
MDL Number: MFCD00001043
Molecular Formula: C6H4BrI
Molecular Weight: 282.91
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 120
Melting point (°C): -9
Flash Point (°C): 110
Specific Gravity (20/20): 2.23


Other Product Names / Synonyms: 3-Bromoiodobenzene

Formula: C₆H₄BrI
Boiling Pt: 120 °C
Melting Pt: –9 °C
Density: 2.23 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00001043
CAS Number: 591-18-4
EINECS: 209-703-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a273dc48c24fa8a4c683be448b158fecbe8096d7&vid=1995e76313d2b9e54bc75a24a2782eba4ff61408&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR