Butylene di(acetate) ≥98.0%

Supplier: TCI America

B1232-25G B1232-500G
TCB1232-025GEA 26.86 CAD
TCB1232-025G TCB1232-500G
Butylene di(acetate) ≥98.0%
Butylene di(acetate)

CAS Number: 628-67-1
MDL Number: MFCD00014984
Molecular Formula: C8H14O4
Molecular Weight: 174.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 230
Freezing point (°C): 13
Specific Gravity (20/20): 1.05


Other Product Names / Synonyms: 1,4-Butanediol Diacetate, Tetramethylene Diacetate

Formula: C₈H₁₄O₄
Boiling Pt: 230 °C
Density: 1.05 (20 °C)
MDL Number: MFCD00014984
CAS Number: 628-67-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4efac4e29f5b99d2e7d2db169083340f76668f7&vid=f4efac4e29f5b99d2e7d2db169083340f76668f7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR