N-Isopropylbenzylamine ≥98.0%

Supplier: TCI America
Warning

Synonyms: N-Benzyl-N-isopropylamine, N-Benzylisopropylamine

B1216-25ML B1216-500ML
TCB1216-025MLEA 127.98 CAD
TCB1216-025ML TCB1216-500ML
N-Isopropylbenzylamine ≥98.0%
N-Isopropylbenzylamine

CAS Number: 102-97-6
MDL Number: MFCD00008863
Molecular Formula: C10H15N
Molecular Weight: 149.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 103
Flash Point (°C): 72
Specific Gravity (20/20): 0.91


Other Product Names / Synonyms: N-Benzylisopropylamine

Formula: C₆H₅CH₂N(CH₃)(CH₂CH₃)
MW: 149.24 g/mol
Boiling Pt: 200
Melting Pt: 0
Density: 0.91 (20 °C)
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00008863
CAS Number: 102-97-6
EINECS: 203-067-6
UN: 2735
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5da93815ab470cdcd203454da239e592ffeed8ce&vid=5da93815ab470cdcd203454da239e592ffeed8ce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR