Bis(2-chloroethyl)ammonium chloride 99,0

Supplier: TCI America

Synonyms: Bis(2-chloroethyl)amine hydrochloride

B1210-25G B1210-500G B1210-100G
TCB1210-025GEA 44.24 CAD
TCB1210-025G TCB1210-500G TCB1210-100G
Bis(2-chloroethyl)ammonium chloride 99,0
Bis(2-chloroethyl)ammonium chloride

CAS Number: 821-48-7
MDL Number: MFCD00012515
Molecular Formula: C4H9Cl2N
Molecular Weight: 178.48
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 213


Other Product Names / Synonyms: 2,2'-Dichlorodiethylamine Hydrochloride

Formula: C₄H₁₀Cl₃N
MW: 178.49 g/mol
Melting Pt: 213 °C
MDL Number: MFCD00012515
CAS Number: 821-48-7
EINECS: 212-479-5
UN: 2923
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=661608eaeef9b94ad821835b5077bb360fb11f66&vid=661608eaeef9b94ad821835b5077bb360fb11f66&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR