N-Benzylpyridin-2-amine 98,0

Supplier: TCI America

Synonyms: 2-(Benzylamino)pyridine

B1209-25G B1209-500G
TCB1209-025GEA 69.13 CAD
TCB1209-025G TCB1209-500G
N-Benzylpyridin-2-amine 98,0
N-Benzylpyridin-2-amine

CAS Number: 6935-27-9
MDL Number: MFCD00006251
Molecular Formula: C12H12N2
Molecular Weight: 184.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 182
Melting point (°C): 95
Flash Point (°C): 153


Other Product Names / Synonyms: N-(2-Pyridyl)benzylamine

Formula: C₁₂H₁₂N₂
MW: 184.24 g/mol
Boiling Pt: 182 °C
Melting Pt: 95 °C
Flash Pt: 153 °C
MDL Number: MFCD00006251
CAS Number: 6935-27-9
EINECS: 230-060-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31f2e92b0b461a6e016eb8e5d1bddf52b8e56c0a&vid=31f2e92b0b461a6e016eb8e5d1bddf52b8e56c0a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR