α,α-Dimethoxytoluene ≥98.0%
Supplier: TCI America
|
Warning
|
B1197-25ML
B1197-500ML
B1197-100ML
TCB1197-025MLEA
41.81
CAD
TCB1197-025ML
TCB1197-500ML
TCB1197-100ML
α,α-Dimethoxytoluene ≥98.0%
α,α-Dimethoxytoluene
CAS Number: 1125-88-8
MDL Number: MFCD00008491
Molecular Formula: C9H12O2
Molecular Weight: 152.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 196
Flash Point (°C): 71
Specific Gravity (20/20): 1.02
Product Categories: Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: alpha,alpha-Dimethoxytoluene
Formula:
C₉H₁₂O₂ Boiling Pt: 196 °C Density: 1.02 (20 °C) Flash Pt: 71 °C |
MDL Number:
MFCD00008491 CAS Number: 1125-88-8 EINECS: 214-413-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...