5-Bromovanillin ≥95.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
B1172-25G
B1172-5G
TCB1172-025GEA
101.12
CAD
TCB1172-025G
TCB1172-5G
5-Bromovanillin ≥95.0% (by titrimetric analysis)
5-Bromovanillin
CAS Number: 2973-76-4
MDL Number: MFCD00006940
Molecular Formula: C8H7BrO3
Molecular Weight: 231.05
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 161
Other Product Names / Synonyms: 5-Bromo-4-hydroxy-3-methoxybenzaldehyde
Formula:
C₈H₇BrO₃ Boiling Pt: 314 °C (1013 hPa) Melting Pt: 161 °C |
MDL Number:
MFCD00006940 CAS Number: 2973-76-4 EINECS: 221-016-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...