2-Bromo-1,4-difluorobenzene ≥98.0%

Supplier: TCI America
Warning

B1166-25G B1166-5G
TCB1166-025GEA 112.34 CAD
TCB1166-025G TCB1166-005G
2-Bromo-1,4-difluorobenzene ≥98.0%
2-Bromo-1,4-difluorobenzene

CAS Number: 399-94-0
MDL Number: MFCD00000345
Molecular Formula: C6H3BrF2
Molecular Weight: 192.99
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 58
Flash Point (°C): 65
Specific Gravity (20/20): 1.72

Formula: C₆H₃BrF₂
Boiling Pt: 58 °C
Melting Pt: –32 °C
Density: 1.72 (20 °C)
Flash Pt: 65 °C
MDL Number: MFCD00000345
CAS Number: 399-94-0
EINECS: 206-920-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3be260075cd13c178bccc35e7325613e906447d&vid=fd39f3e93c7673beb7c1f8c034070949fdea03b3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR