4-Benzoylbenzoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B1164-25G B1164-5G
TCB1164-25GEA 386.78 CAD
TCB1164-25G TCB1164-005G
4-Benzoylbenzoic acid ≥98.0% (by titrimetric analysis)
4-Benzoylbenzoic acid

CAS Number: 611-95-0
MDL Number: MFCD00002560
Molecular Formula: C14H10O3
Molecular Weight: 226.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 196


Product Categories: Materials Chemistry, Polymerization Initiators
Other Product Names / Synonyms: Benzophenone-4-carboxylic Acid

Formula: C₁₄H₁₀O₃
Boiling Pt: 416 °C (1013 hPa)
Melting Pt: 196 °C
MDL Number: MFCD00002560
CAS Number: 611-95-0
EINECS: 210-286-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3187189e8093aff72fe8cacfb7cca39d6e3e3b15&vid=3187189e8093aff72fe8cacfb7cca39d6e3e3b15&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR