Benzophenone hydrazone ≥98.0% (by HPLC)

Supplier: TCI America

B1163-500G B1163-25G
TCB1163-500GEA 213.3 CAD
TCB1163-500G TCB1163-025G
Benzophenone hydrazone ≥98.0% (by HPLC)
Benzophenone hydrazone

CAS Number: 5350-57-2
MDL Number: MFCD00007624
Molecular Formula: C13H12N2
Molecular Weight: 196.25
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Boiling point (°C): 230
Melting point (°C): 99
Flash Point (°C): 156

Formula: C₁₃H₁₂N₂
MW: 196.25 g/mol
Boiling Pt: 230 °C
Melting Pt: 99 °C
Flash Pt: 156 °C
MDL Number: MFCD00007624
CAS Number: 5350-57-2
EINECS: 226-321-8
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6d5050bc6288590c75ebfd40f156328573b9493&vid=e6d5050bc6288590c75ebfd40f156328573b9493&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR