(2R,3R)-(-)-2,3-Butanediol ≥97.0%

Supplier: TCI America

B1161-5G B1161-1G
TCB1161-005GEA 302.25 CAD
TCB1161-005G TCB1161-001G
(2R,3R)-(-)-2,3-Butanediol ≥97.0%
(2R,3R)-(-)-2,3-Butanediol

CAS Number: 24347-58-8
MDL Number: MFCD00064267
Molecular Formula: C4H10O2
Molecular Weight: 90.12
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 80
Flash Point (°C): 85
Freezing point (°C): 17
Specific Gravity (20/20): 0.99
Specific rotation [a]20/D: -13 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Analytical Science, e.e. / Absolute Configuration Determination (NMR)
Other Product Names / Synonyms: (R,R)-(-)-2,3-Butylene Glycol, (R,R)-(-)-2,3-Dihydroxybutane

Formula: C₄H₁₀O₂
Boiling Pt: 80 °C
Melting Pt: 16 °C
Density: 0.99 (20 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00064267
CAS Number: 24347-58-8
EINECS: 246-186-9
Merck Index: 13,01567
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=deed47ce80da158ba72a1104db79df4cef9bb4ac&vid=deed47ce80da158ba72a1104db79df4cef9bb4ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR