Benzo-15-crown-5 (benzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥98.0%
Supplier: TCI America
Synonyms:
Benzo-15-crown-5
B1154-1G
B1154-5G
TCB1154-1GEA
107.44
CAD
TCB1154-1G
TCB1154-005G
Benzo-15-crown-5 (benzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥98.0%
Benzo-15-crown-5 (Benzo-1,4,7,10,13-pentaoxacyclopentadecane)
CAS Number: 14098-44-3
MDL Number: MFCD00005945
Molecular Formula: C14H20O5
Molecular Weight: 268.31
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 80
Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: 2,3-Benzo-1,4,7,10,13-pentaoxacyclopentadec-2-ene
Formula:
C₁₄H₂₀O₅ MW: 268.31 g/mol Melting Pt: 80 °C |
MDL Number:
MFCD00005945 CAS Number: 14098-44-3 EINECS: 237-947-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...