1,3-Bis(diphenylphosphino)propane ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
Propylenebis(diphenylphosphine), Propane-1,3-diylbis(diphenylphosphane), Propane-1,3-diylbis(diphenylphosphine), 1,3-Bis(diphenylphosphino)propane, DPPP, Bis(1,2-diphenylphosphino)propane
CAS Number: 6737-42-4
MDL Number: MFCD00003050
Molecular Formula: C27H26P2
Molecular Weight: 412.45
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 63
Flash Point (°C): 235
Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, C-H Bond Activation Reaction, Ligands, Ligands ([2+2+2]Cycloaddition Reaction)
Other Product Names / Synonyms: Trimethylenebis(diphenylphosphine)
Formula:
C₂₇H₂₆P₂ MW: 412.45 g/mol Melting Pt: 63 °C Flash Pt: 235 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003050 CAS Number: 6737-42-4 EINECS: 229-791-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...