5-Bromothiophene-2-carbaldehyde ≥97.0%

Supplier: TCI America

B1135-5G B1135-25G
TCB1135-5GEA 55.3 CAD
TCB1135-5G TCB1135-25G
5-Bromothiophene-2-carbaldehyde ≥97.0%
5-Bromothiophene-2-carbaldehyde

CAS Number: 4701-17-1
MDL Number: MFCD00005432
Molecular Formula: C5H3BrOS
Molecular Weight: 191.04
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 115
Flash Point (°C): 98
Freezing point (°C): 15
Specific Gravity (20/20): 1.77
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 5-Bromo-2-formylthiophene

Formula: C₅H₃BrOS
Boiling Pt: 115 °C
Density: 1.77 (20 °C)
Flash Pt: 98 °C
MDL Number: MFCD00005432
CAS Number: 4701-17-1
EINECS: 225-176-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76050205cd53586b900a1a210d7e5f7fd85ff937&vid=76050205cd53586b900a1a210d7e5f7fd85ff937&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR