2-(2-Bromoethyl)-1,3-dioxolane ≥90.0%
Supplier: TCI America
B1132-25G
B1132-100G
TCB1132-025GEA
160.37
CAD
TCB1132-025G
76219-332
2-(2-Bromoethyl)-1,3-dioxolane ≥90.0%
2-(2-Bromoethyl)-1,3-dioxolane
CAS Number: 18742-02-4
MDL Number: MFCD00003216
Molecular Formula: C5H9BrO2
Molecular Weight: 181.03
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 73
Flash Point (°C): 65
Specific Gravity (20/20): 1.53
Storage Temperature: <0°C
Other Product Names / Synonyms: 3-Bromopropionaldehyde Ethylene Acetal
Formula:
C₅H₉BrO₂ Boiling Pt: 73 °C Density: 1.53 (20 °C) Flash Pt: 65 °C |
MDL Number:
MFCD00003216 CAS Number: 18742-02-4 EINECS: 242-551-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...