2-Benzylaminoethanol ≥95.0%

Supplier: TCI America
Warning

Synonyms: N-Benzylethanolamine, 2-(Benzylamino)ethanol

B1130-25ML B1130-500ML
TCB1130-025MLEA 70.34 CAD
TCB1130-025ML TCB1130-500ML
2-Benzylaminoethanol ≥95.0%
2-Benzylaminoethanol

CAS Number: 104-63-2
MDL Number: MFCD00002840
Molecular Formula: C9H13NO
Molecular Weight: 151.21
Purity/Analysis Method: >95.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 280
Flash Point (°C): 130
Specific Gravity (20/20): 1.06


Other Product Names / Synonyms: 2-Benzylaminoethanol, N-(2-Hydroxyethyl)benzylamine

Formula: C₆H₅CH₂NHCH₂CH₂OH
MW: 151.21 g/mol
Boiling Pt: 280 °C
Density: 1.06 (20 °C)
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00002840
CAS Number: 104-63-2
EINECS: 203-221-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d04f8caea587d9d7a5cf7943cbf758e3f53cd92f&vid=d04f8caea587d9d7a5cf7943cbf758e3f53cd92f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR