1-Bromo-2-iodobenzene ≥98.0% stabilized
Supplier: TCI America
|
Warning
|
Synonyms:
2-Bromo-1-iodobenzene, 2-Bromoiodobenzene
Stabilization:
Stabilised with Stabilised
B1126-5G
B1126-250G
B1126-25G
TCB1126-005GEA
44.24
CAD
TCB1126-005G
TCB1126-250G
TCB1126-025G
1-Bromo-2-iodobenzene ≥98.0% stabilized
1-Bromo-2-iodobenzene
(stabilized with Copper chip)
CAS Number: 583-55-1
MDL Number: MFCD00001030
Molecular Formula: C6H4BrI
Molecular Weight: 282.91
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 120
Flash Point (°C): 113
Freezing point (°C): 7
Specific Gravity (20/20): 2.26
Other Product Names / Synonyms: 2-Bromoiodobenzene
Formula:
C₆H₄BrI MW: 282.91 g/mol Boiling Pt: 120 °C Density: 2.26 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00001030 CAS Number: 583-55-1 EINECS: 209-508-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...