4-Benzyloxyphenol ≥99.0%

Supplier: TCI America

Synonyms: Monobenzone4-Hydroxyphenyl benzyl etherHydroquinone monobenzyl ether4-(Benzyloxy)phenol

B1104-25G B1104-500G B1104-100G
TCB1104-025GEA 63.2 CAD
TCB1104-025G TCB1104-500G TCB1104-100G
4-Benzyloxyphenol ≥99.0%
4-Benzyloxyphenol

CAS Number: 103-16-2
MDL Number: MFCD00002333
Molecular Formula: C13H12O2
Molecular Weight: 200.24
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 121


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: Hydroquinone Monobenzyl Ether

Formula: C₁₃H₁₂O₂
MW: 200.24 g/mol
Boiling Pt: 328
Melting Pt: 121 °C
Density: 1.26 g/cm³ (20 °C)
MDL Number: MFCD00002333
CAS Number: 103-16-2
EINECS: 203-083-3
Merck Index: 12,06331
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a6ab75fca7252429e61470f19592d362e786610&vid=1a6ab75fca7252429e61470f19592d362e786610&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR