Pentyl benzoate ≥98.0%

Supplier: TCI America

B1095-25ML B1095-500ML B1095-100ML
TCB1095-025MLEA 33.1 CAD
TCB1095-025ML TCB1095-500ML TCB1095-100ML
Pentyl benzoate ≥98.0%
Pentyl benzoate

CAS Number: 2049-96-9
MDL Number: MFCD00048843
Molecular Formula: C12H16O2
Molecular Weight: 192.26
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 260
Flash Point (°C): 130
Specific Gravity (20/20): 0.99


Other Product Names / Synonyms: Benzoic Acid Amyl Ester, Pentyl Benzoate

Formula: C₁₂H₁₆O₂
Boiling Pt: 260 °C
Density: 0.99 (20 °C)
Flash Pt: 130 °C
MDL Number: MFCD00048843
CAS Number: 2049-96-9
EINECS: 218-077-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b34af5f6eca2d0cfc6c7e9cd4e23f2e10a4e2668&vid=1ba2755d83a099c0719213a7af3847885fb1fb34&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR