6-Benzylaminopurine ≥99.0%
Supplier: TCI America
Synonyms:
6-BAP, 6-Benzyladenine, N6-Benzyladenine, Benzyl(purin-6-yl)amine
B1088-25G
B1088-5G
TCB1088-025GEA
260.7
CAD
TCB1088-025G
TCB1088-005G
6-Benzylaminopurine ≥99.0%
6-Benzylaminopurine
CAS Number: 1214-39-7
MDL Number: MFCD00005572
Molecular Formula: C12H11N5
Molecular Weight: 225.26
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Melting point (°C): 234
Product Categories: Bioscience, Cytokinins, Nucleobases and Their Analogs
Other Product Names / Synonyms: 6-Benzylaminopurine
Formula:
C₁₂H₁₁N₅ MW: 225.25 g/mol Melting Pt: 234 °C |
MDL Number:
MFCD00005572 CAS Number: 1214-39-7 EINECS: 214-927-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...