Hexyl benzoate ≥98.0%

Supplier: TCI America

Synonyms: Benzoic acid hexyl ester

B1076-500ML B1076-25ML
TCB1076-500MLEA 101.12 CAD
TCB1076-500ML TCB1076-025ML
Hexyl benzoate ≥98.0%
Hexyl benzoate

CAS Number: 6789-88-4
MDL Number: MFCD00051714
Molecular Formula: C13H18O2
Molecular Weight: 206.29
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 272
Flash Point (°C): 148
Specific Gravity (20/20): 0.98


Other Product Names / Synonyms: Benzoic Acid Hexyl Ester

Formula: C₁₃H₁₈O₂
MW: 206.28 g/mol
Boiling Pt: 272 °C
Density: 0.98 (20 °C)
Flash Pt: 148 °C
MDL Number: MFCD00051714
CAS Number: 6789-88-4
EINECS: 229-856-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6728877c5b01c60d4928e91527c1e5133666c4c&vid=6b115a2df4a749be55c532747630d7391b214171&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR