4-tert-Butylcyclohexanone ≥97.0%

Supplier: TCI America

B1074-25G B1074-500G B1074-100G
TCB1074-025GEA 126.4 CAD
TCB1074-025G TCB1074-500G TCB1074-100G
4-tert-Butylcyclohexanone ≥97.0%
4-tert-Butylcyclohexanone

CAS Number: 98-53-3
MDL Number: MFCD00001642
Molecular Formula: C10H18O
Molecular Weight: 154.25
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 116
Melting point (°C): 50
Flash Point (°C): 95

Formula: C₁₀H₁₈O
Boiling Pt: 116 °C
Melting Pt: 50 °C
Density: 0.893 g/cm³ (50 °C)
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00001642
CAS Number: 98-53-3
EINECS: 202-678-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31b89123d638404f78a1698934de0a8f61e5b5e5&vid=31b89123d638404f78a1698934de0a8f61e5b5e5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR