Vinyl benzoate ≥99.0% stabilized

Supplier: TCI America


Stabilization: Stabilised with Stabilised

B1032-500ML B1032-25ML
TCB1032-500MLEA 311.26 CAD
TCB1032-500ML TCB1032-025ML
Vinyl benzoate ≥99.0% stabilized
Vinyl benzoate

(stabilized with MEHQ)
CAS Number: 769-78-8
MDL Number: MFCD00048141
Molecular Formula: C9H8O2
Molecular Weight: 148.16
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 203
Melting point (°C): -32
Flash Point (°C): 82
Specific Gravity (20/20): 1.07


Other Product Names / Synonyms: Benzoic Acid Vinyl Ester

Formula: C₉H₈O₂
Boiling Pt: 203 °C
Melting Pt: –32 °C
Density: 1.07 (20 °C)
Flash Pt: 82 °C
MDL Number: MFCD00048141
CAS Number: 769-78-8
EINECS: 212-214-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23340a90fa4d0acddf72b8172f9fbae42bcbf859&vid=23340a90fa4d0acddf72b8172f9fbae42bcbf859&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR