1,8-Bis(dimethylamino)naphthalene ≥98.0%
Supplier: TCI America
Synonyms:
N1,N1,N8,N8-Tetramethylnaphthalene-1,8-diamine
[for Dehydrohalogenation]
CAS Number: 20734-58-1
MDL Number: MFCD00003920
Molecular Formula: C14H18N2
Molecular Weight: 214.31
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 145
Melting point (°C): 48
Flash Point (°C): 113
Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts
Other Product Names / Synonyms: N,N,N',N'-Tetramethyl-1,8-diaminonaphthalene
Formula:
C₁₄H₁₈N₂ MW: 214.31 g/mol Boiling Pt: 145 °C Melting Pt: 48 °C Flash Pt: 113 °C |
MDL Number:
MFCD00003920 CAS Number: 20734-58-1 EINECS: 244-001-6 UN: 3263 ADR: 8 III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...