4-Butylbenzaldehyde ≥95.0%
Supplier: TCI America
|
Warning
|
B1014-25ML
B1014-5ML
TCB1014-25MLEA
70.71
CAD
TCB1014-25ML
TCB1014-005ML
4-Butylbenzaldehyde ≥95.0%
4-Butylbenzaldehyde
CAS Number: 1200-14-2
MDL Number: MFCD00040751
Molecular Formula: C11H14O
Molecular Weight: 162.23
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 165
Flash Point (°C): 128
Specific Gravity (20/20): 0.97
Formula:
C₁₁H₁₄O Boiling Pt: 165 °C Melting Pt: 43 °C Density: 0.97 (20 °C) Flash Pt: 128 °C |
MDL Number:
MFCD00040751 CAS Number: 1200-14-2 EINECS: 214-851-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...