2-Bromo-p-xylene ≥99.0%

Supplier: TCI America
Warning

B0997-25G B0997-500G
TCB0997-025GEA 24.76 CAD
TCB0997-025G TCB0997-500G
2-Bromo-p-xylene ≥99.0%
2-Bromo-p-xylene

CAS Number: 553-94-6
MDL Number: MFCD00000074
Molecular Formula: C8H9Br
Molecular Weight: 185.06
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 201
Flash Point (°C): 79
Freezing point (°C): 7
Specific Gravity (20/20): 1.36


Other Product Names / Synonyms: 2-Bromo-1,4-dimethylbenzene

Formula: C₈H₉Br
Boiling Pt: 201 °C
Melting Pt: 8.9 °C
Density: 1.36 (20 °C)
Flash Pt: 79 °C
MDL Number: MFCD00000074
CAS Number: 553-94-6
EINECS: 209-055-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec0971bdcf0e90d8d6e45a6b1d4ce4ff9fde9831&vid=7423cee95973a0e59888b14025816e35d5f3ae39&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR