4-Bromo-1,2-dichlorobenzene ≥98.0%

Supplier: TCI America
Warning

Synonyms: 1-Bromo-3,4-dichlorobenzene

B0983-25G B0983-500G B0983-100G
TCB0983-025GEA 69.52 CAD
TCB0983-025G TCB0983-500G 76219-328
4-Bromo-1,2-dichlorobenzene ≥98.0%
4-Bromo-1,2-dichlorobenzene

CAS Number: 18282-59-2
MDL Number: MFCD00040849
Molecular Formula: C6H3BrCl2
Molecular Weight: 225.89
Purity/Analysis Method: >98.0% (GC)
Form: Lump
Boiling point (°C): 124
Flash Point (°C): 87
Freezing point (°C): 23


Other Product Names / Synonyms: 4-Bromo-1,2-dichlorobenzene

Formula: C₆H₃BrCl₂
MW: 225.9 g/mol
Boiling Pt: 124 °C
Melting Pt: 24.5
Density: 1.781 g/cm³ (20 °C)
Flash Pt: 87 °C
MDL Number: MFCD00040849
CAS Number: 18282-59-2
EINECS: 242-160-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=453d4dce7d0a49ab4d968e33e72dac67c5e524c6&vid=1429d58aadb86e97b9573b3c495652ad1edf02a2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR