1-Bromo-2,6-dichlorobenzene ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
2-Bromo-1,3-dichlorobenzene
B0982-25G
B0982-5G
TCB0982-025GEA
113.76
CAD
TCB0982-025G
TCB0982-5G
1-Bromo-2,6-dichlorobenzene ≥98.0%
1-Bromo-2,6-dichlorobenzene
CAS Number: 19393-92-1
MDL Number: MFCD00000574
Molecular Formula: C6H3BrCl2
Molecular Weight: 225.89
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 242
Melting point (°C): 66
Formula:
C₆H₃BrCl₂ MW: 225.9 g/mol Boiling Pt: 242 °C Melting Pt: 66 °C Density: 2.077 g/cm³ (20 °C) |
MDL Number:
MFCD00000574 CAS Number: 19393-92-1 EINECS: 243-018-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...