1-Bromo-2,3-dichlorobenzene ≥99.0%

Supplier: TCI America
Warning

B0980-5G B0980-25G
TCB0980-005GEA 28.44 CAD
TCB0980-005G TCB0980-025G
1-Bromo-2,3-dichlorobenzene ≥99.0%
1-Bromo-2,3-dichlorobenzene

CAS Number: 56961-77-4
MDL Number: MFCD00000536
Molecular Formula: C6H3BrCl2
Molecular Weight: 225.89
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 243
Melting point (°C): 60

Formula: C₆H₃BrCl₂
Boiling Pt: 243 °C
Melting Pt: 60 °C
MDL Number: MFCD00000536
CAS Number: 56961-77-4
EINECS: 260-476-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4fdaeb00185b9565fb6141bdaee57bea87046edd&vid=3cec91531832a1e8c80f49d2218608a45a2a8bc0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR