3,3',4,4'-Benzophenonetetracarboxylic dianhydride ≥96.0%

Supplier: TCI America

Synonyms: Benzophenone-3,3',4,4'-tetracarboxylic dianhydride

B0948-25G B0948-500G B0948-100G
TCB0948-025GEA 85.32 CAD
TCB0948-025G TCB0948-500G TCB0948-100G
3,3',4,4'-Benzophenonetetracarboxylic dianhydride ≥96.0%
3,3',4,4'-Benzophenonetetracarboxylic dianhydride

CAS Number: 2421-28-5
MDL Number: MFCD00005923
Molecular Formula: C17H6O7
Molecular Weight: 322.23
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Melting point (°C): 226
Flash Point (°C): 324
Lambda max.: 580 nm (CH3CN)


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials, Benzophenones (for High-Performance Polymer Research)
Other Product Names / Synonyms: 4,4'-Carbonyldiphthalic Anhydride

Formula: C₁₇H₆O₇
MW: 322.23 g/mol
Melting Pt: 226 °C
Flash Pt: 324 °C
MDL Number: MFCD00005923
CAS Number: 2421-28-5
EINECS: 219-348-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8af0a3d2f70ede3bf0248867363c5f85ff571e6c&vid=8af0a3d2f70ede3bf0248867363c5f85ff571e6c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR