1-Butanol ≥99.0%

Supplier: TCI America

Synonyms: n-ButanolPropyl carbinol1-Butyl alcoholButyl Alcohol1-Hydroxy butanen-Butyl Alcohol

B0944-500ML
TCB0944-500MLEA 71.97 CAD
TCB0944-500ML
1-Butanol ≥99.0%
1-Butanol

[for HPLC Solvent]
CAS Number: 71-36-3
MDL Number: MFCD00002964
Molecular Formula: C4H10O
Molecular Weight: 74.12
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 118
Melting point (°C): -90
Flash Point (°C): 37
Specific Gravity (20/20): 0.81


Product Categories: Analytical Science, Solvents for HPLC
Other Product Names / Synonyms: Butyl Alcohol

Formula: H₃C(CH₂)₂CH₂OH
MW: 74.12 g/mol
Boiling Pt: 118 °C
Melting Pt: –90 °C
Density: 0.81 (20 °C)
Flash Pt: 37 °C
MDL Number: MFCD00002964
CAS Number: 71-36-3
UN: 1120
ADR: 3,III
Merck Index: 13,01539
https://ca.vwr.com/stibo/web/std.lang.all/28/23/28932823.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR