Benzoin isopropyl ether ≥99.0%

Supplier: TCI America

B0942-25G B0942-500G
TCB0942-025GEA 75.57 CAD
TCB0942-025G TCB0942-500G
Benzoin isopropyl ether ≥99.0%
Benzoin isopropyl ether

CAS Number: 6652-28-4
MDL Number: MFCD00015025
Molecular Formula: C17H18O2
Molecular Weight: 254.33
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 180
Melting point (°C): 79
Flash Point (°C): 161


Product Categories: Materials Chemistry, Polymerization Initiators
Other Product Names / Synonyms: 2-Isopropoxy-2-phenylacetophenone

Formula: C₁₇H₁₈O₂
Boiling Pt: 180 °C
Melting Pt: 79 °C
Flash Pt: 161 °C
MDL Number: MFCD00015025
CAS Number: 6652-28-4
EINECS: 229-677-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=638ff9d1c268adee2c3c047e2e85ae20ca37434b&vid=638ff9d1c268adee2c3c047e2e85ae20ca37434b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR