1-Bromoundecane ≥98.0%
Supplier: TCI America
Synonyms:
1-Undecyl bromide
B0934-500ML
B0934-25ML
TCB0934-500MLEA
482.31
CAD
TCB0934-500ML
TCB0934-025ML
1-Bromoundecane ≥98.0%
1-Bromoundecane
CAS Number: 693-67-4
MDL Number: MFCD00000223
Molecular Formula: C11H23Br
Molecular Weight: 235.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 138
Melting point (°C): -9
Flash Point (°C): 113
Specific Gravity (20/20): 1.06
Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: Hendecyl Bromide, Undecyl Bromide
Formula:
BrH₂C(CH₂)₉CH₃ MW: 235.21 g/mol Boiling Pt: 138 °C Melting Pt: –9 °C Density: 1.06 (20 °C) Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000223 CAS Number: 693-67-4 EINECS: 211-757-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...