1-Bromononane ≥98.0%

Supplier: TCI America

B0933-25ML B0933-500ML B0933-100ML
TCB0933-025MLEA 66.36 CAD
TCB0933-025ML TCB0933-500ML 76219-326
1-Bromononane ≥98.0%
1-Bromononane

CAS Number: 693-58-3
MDL Number: MFCD00000278
Molecular Formula: C9H19Br
Molecular Weight: 207.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 201
Flash Point (°C): 90
Specific Gravity (20/20): 1.09


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: Nonyl Bromide

Formula: BrH₂C(CH₂)₇CH₃
Boiling Pt: 201 °C
Melting Pt: –29 °C
Density: 1.09 (20 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00000278
CAS Number: 693-58-3
EINECS: 211-755-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25c0fdf7d320a209433e6ce82b5d592485a80494&vid=807dbbad41a8b4bc0d07e380a177a58a49d87752&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR