4-Butoxyphenol ≥97.0%

Supplier: TCI America

Synonyms: Hydroquinone monobutyl ether

B0923-500G B0923-25G
TCB0923-500GEA 1295.92 CAD
TCB0923-500G TCB0923-25G
4-Butoxyphenol ≥97.0%
4-Butoxyphenol

CAS Number: 122-94-1
MDL Number: MFCD00002336
Molecular Formula: C10H14O2
Molecular Weight: 166.22
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 165
Melting point (°C): 66

Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
Boiling Pt: 165 °C
Melting Pt: 66 °C
MDL Number: MFCD00002336
CAS Number: 122-94-1
EINECS: 204-583-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7ed1e0a9bae5fcfd9d435d32ac4f4b7f40cbdad&vid=e7ed1e0a9bae5fcfd9d435d32ac4f4b7f40cbdad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR