2,1,3-Benzothiadiazole ≥99.0%

Supplier: TCI America

B0921-10G B0921-25G
TCB0921-010GEA 82.16 CAD
TCB0921-010G TCB0921-25G
2,1,3-Benzothiadiazole ≥99.0%
2,1,3-Benzothiadiazole

CAS Number: 273-13-2
MDL Number: MFCD00005809
Molecular Formula: C6H4N2S
Molecular Weight: 136.17
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 64
Melting point (°C): 44
Flash Point (°C): 95


Product Categories: Materials Chemistry, Conducting Polymer Research Reagents
Other Product Names / Synonyms: Piazthiole

Formula: C₆H₄N₂S
MW: 136.18 g/mol
Boiling Pt: 64 °C
Melting Pt: 44 °C
Flash Pt: 95 °C
MDL Number: MFCD00005809
CAS Number: 273-13-2
EINECS: 205-985-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=888a02729027d178c74578e4b02369ee846ca0a2&vid=888a02729027d178c74578e4b02369ee846ca0a2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR