2-[Bis(2-hydroxyethyl)amino]ethanesulfonic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B0909-25G B0909-500G
TCB0909-25GEA 58.46 CAD
TCB0909-25G TCB0909-500G
2-[Bis(2-hydroxyethyl)amino]ethanesulfonic acid ≥98.0% (by titrimetric analysis)
2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid

[Good's buffer component for biological research]
CAS Number: 10191-18-1
MDL Number: MFCD00007533
Molecular Formula: C6H15NO5S
Molecular Weight: 213.25
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 155


Product Categories: Bioscience, Good's Buffering Agents
Other Product Names / Synonyms: BES

Formula: C₆H₁₅NO₅S
Boiling Pt: 405 °C (1013 hPa)
Melting Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00007533
CAS Number: 10191-18-1
EINECS: 233-465-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f8cf2e325ff4de0f5a283ce13b3d389d367d7c9&vid=370ad96b69774a291df234349a5ee866ff61a165&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR