Bicyclohexyl ≥99.0%

Supplier: TCI America

B0902-500ML B0902-25ML B0902-100ML
TCB0902-500MLEA 808.33 CAD
TCB0902-500ML TCB0902-025ML TCB0902-100ML
Bicyclohexyl ≥99.0%
Bicyclohexyl

CAS Number: 92-51-3
MDL Number: MFCD00003815
Molecular Formula: C12H22
Molecular Weight: 166.31
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 239
Flash Point (°C): 92
Freezing point (°C): 3
Specific Gravity (20/20): 0.89


Other Product Names / Synonyms: Dicyclohexyl

Formula: C₁₂H₂₂
Boiling Pt: 239 °C
Density: 0.89 (20 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00003815
CAS Number: 92-51-3
EINECS: 202-161-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb010a1dfb6c139901043e1827c5565efce0eee7&vid=cb010a1dfb6c139901043e1827c5565efce0eee7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR