Benzoyl ferrocene ≥98.0%

Supplier: TCI America

Synonyms: Benzoylferrocene

B0886-5G B0886-25G
TCB0886-005GEA 91.64 CAD
TCB0886-005G TCB0886-025G
Benzoyl ferrocene ≥98.0%
Benzoylferrocene

CAS Number: 1272-44-2
MDL Number: MFCD00001431
Molecular Formula: C17H14FeO
Molecular Weight: 290.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 110

Formula: C₁₇H₁₄FeO
Melting Pt: 110 °C
MDL Number: MFCD00001431
CAS Number: 1272-44-2
EINECS: 215-054-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=08491c80d87208e25eec9eccc7673e20f960aef9&vid=08491c80d87208e25eec9eccc7673e20f960aef9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR