9-Bromoanthracene ≥95.0%
Supplier: TCI America
B0872-5G
B0872-25G
TCB0872-5GEA
34.76
CAD
TCB0872-5G
TCB0872-025G
9-Bromoanthracene ≥95.0%
9-Bromoanthracene
CAS Number: 1564-64-3
MDL Number: MFCD00001243
Molecular Formula: C14H9Br
Molecular Weight: 257.13
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 101
Formula:
C₁₄H₉Br Boiling Pt: 359 °C (1013 hPa) Melting Pt: 101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001243 CAS Number: 1564-64-3 EINECS: 216-359-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...